accessibility menu, dialog, popup

UserName

CAS RN 1826-28-4

IUPAC Name: (2Z)-2-[nitroso(phenyl)methylidene]-1,2-dihydropyridine
Synonyms: (2Z)-2-[nitroso(phenyl)methylidene]-1H-pyridine
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: WBSIGZZXFQOEJK-QXMHVHEDSA-N
SMILES: O=N\C(=C1/NC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results