missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-O-Acetyl-3,4,6-tri-O-benzyl-1-thio-beta-D-galactopyranoside, TCI America™

Catalog No. P20781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20781G Supplier TCI America Supplier No. P20781G
Only null left

Chemical Identifiers

CAS 183875-28-7
Molecular Formula C35H36O6S
Molecular Weight (g/mol) 584.727
InChI Key IKFDEPUTIUTSLC-NVCPMKERSA-N
Synonym Gal[2Ac,346Bn]-beta-SPh
PubChem CID 124527361
IUPAC Name [(2S,3R,4S,5S,6R)-4,5-bis(phenylmethoxy)-6-(phenylmethoxymethyl)-2-phenylsulfanyloxan-3-yl] acetate
SMILES CC(=O)OC1C(C(C(OC1SC2=CC=CC=C2)COCC3=CC=CC=C3)OCC4=CC=CC=C4)OCC5=CC=CC=C5

Specifications

Melting Point 111°C
Quantity 1 g
Formula Weight 584.73
Chemical Name or Material Phenyl 2-O-Acetyl-3,4,6-tri-O-benzyl-1-thio-beta-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator