accessibility menu, dialog, popup

UserName

CAS RN 183875-28-7

CH3OOOSOOO

CAS RN 183875-28-7

IUPAC Name: 4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]-2-(phenylsulfanyl)oxan-3-yl acetate
Synonyms: 4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]-2-(phenylsulfanyl)oxan-3-yl acetate
Molecular Weight (g/mol): 584.73
Molecular Formula: C35H36O6S
InChi Key: IKFDEPUTIUTSLC-UHFFFAOYNA-N
SMILES: CC(=O)OC1C(OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1)SC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results