missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,4,6-Tri-O-acetyl-3-O-allyl-1-thio-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. p16801g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P16801G Supplier TCI America Supplier No. P16801G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1820572-28-8
Molecular Formula C21H26O8S
Molecular Weight (g/mol) 438.491
InChI Key AEZBNOXQHIZNTR-IFLJBQAJSA-N
Synonym Gal[246Ac,3All]-beta-SPh
PubChem CID 91659150
IUPAC Name [(2R,3S,4S,5R,6S)-3,5-diacetyloxy-6-phenylsulfanyl-4-prop-2-enoxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OCC=C)OC(=O)C

Specifications

Color White
Quantity 1 g
Formula Weight 438.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Phenyl 2,4,6-Tri-O-acetyl-3-O-allyl-1-thio-beta-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator