accessibility menu, dialog, popup

UserName

CAS RN 1820572-28-8

IUPAC Name: [3,5-bis(acetyloxy)-6-(phenylsulfanyl)-4-(prop-2-en-1-yloxy)oxan-2-yl]methyl acetate
Synonyms: [3,5-bis(acetyloxy)-6-(phenylsulfanyl)-4-(prop-2-en-1-yloxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 438.49
Molecular Formula: C21H26O8S
InChi Key: AEZBNOXQHIZNTR-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(SC2=CC=CC=C2)C(OC(C)=O)C(OCC=C)C1OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results