missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™

Supplier:  TCI America P147625G

 View more versions of this product

Catalog No. P147625G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside
118°C
C20H24O9S
25 g
JCKOUAWEMPKIAT-OBKDMQGPSA-N
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
6364823
≥98.0% (HPLC)
23661-28-1
White
MFCD00135159
2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 1-phenyl-1-thio-, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
440.463
440.46
Crystalline Powder

Chemical Identifiers

23661-28-1
440.463
JCKOUAWEMPKIAT-OBKDMQGPSA-N
6364823
CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C
C20H24O9S
MFCD00135159
2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 1-phenyl-1-thio-, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions