missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside 98.0+%, TCI America™
$274.02 - $941.35
Chemical Identifiers
| CAS | 23661-28-1 |
|---|---|
| Molecular Formula | C20H24O9S |
| Molecular Weight (g/mol) | 440.463 |
| MDL Number | MFCD00135159 |
| InChI Key | JCKOUAWEMPKIAT-OBKDMQGPSA-N |
| Synonym | 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 1-phenyl-1-thio-, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate |
| PubChem CID | 6364823 |
| IUPAC Name | [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate |
| SMILES | CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C |
Chemical Identifiers
| 23661-28-1 | |
| 440.463 | |
| JCKOUAWEMPKIAT-OBKDMQGPSA-N | |
| 6364823 | |
| CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C |
| C20H24O9S | |
| MFCD00135159 | |
| 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 2r,3r,4s,5r,6s-3,4,5-tris acetyloxy-6-phenylsulfanyl oxan-2-yl methyl acetate, 1-phenyl-1-thio-, 1-phenyl-1-thio-, a-d-glucopyranoside tetraacetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate | |
| [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate |
Specifications
| 23661-28-1 | |
| White | |
| MFCD00135159 | |
| 2r,3r,4s,5r,6s-2-acetoxymethyl-6-phenylthio tetrahydro-2h-pyran-3,4,5-triyl triacetate, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-b-d-thioglucopyranoside, phenyl 2,3,4,6-tetra-o-acetyl-1-thio-beta-d-glucopyranoside, 10.14272/jckouawempkiat-obkdmqgpsa-n, 1-phenyl-1-thio-, 1-phenyl-1-thio-, doi:10.14272/jckouawempkiat-obkdmqgpsa-n, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, s-phenyl-1-thio-beta-d-glucopyranoside tetracetate, 1-phenylthio-1-deoxy-beta-d-glucopyranose tetraacetate | |
| CC(=O)OCC1C(C(C(C(O1)SC2=CC=CC=C2)OC(=O)C)OC(=O)C)OC(=O)C | |
| 440.463 | |
| 440.46 | |
| Crystalline Powder |
| 118°C | |
| C20H24O9S | |
| 5 g | |
| JCKOUAWEMPKIAT-OBKDMQGPSA-N | |
| [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-phenylsulfanyloxan-2-yl]methyl acetate | |
| 6364823 | |
| ≥98.0% (HPLC) | |
| Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside |
Safety and Handling
TSCA : No
Recommended Storage : Freezer