missing translation for 'onlineSavingsMsg'
Learn More

Phentolamine Mesylate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015076
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015076 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015076 Supplier Merck Emd Millipore Supplier No. PHR14031G
Phentolamine Mesylate belongs to the class of alpha adrenergic antagonist drugs. It is used in the treatment of pheochromocytoma-related hypertension, erectile dysfunction and in the diagnosis of pheochromocytoma and norepinephrine-related dermal necrosis.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 65-28-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C17H19N3 O · CH3 SO3 H
MDL Number MFCD00134201
Melting Point 180°C to 182°C
Synonym Phentolamine methanesulfonate salt; 2-(N-[m-Hydroxyphenyl]-p-toluidinomethyl)imidazoline
Type Certified Reference Material
Quantity 1 g
Formula Weight 377.46
Grade Certified Reference Material
Show More Show Less