missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyethanol, Reagent, 99%, Spectrum™ Chemical

Catalog No. p-7157976
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605108 120 mL Amber Glass Bottle
18605109 500 mL Amber Glass Bottle
18605110 1 L Amber Glass Bottle
18605111 4 L Amber Glass Bottle
18605112 4 x 4 L Amber Glass Bottle
18605113 20 L Amber Glass Bottle
18605114 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18605108 Supplier Spectrum Chemical Mfg Cor Supplier No. P1077120ML
Only null left

P1077, 122-99-6, C8H10O2

Spectrum™ Chemical Phenoxyethanol, Reagent , is a glycol ether and an organic chemical compound. This colorless oily liquid is often used in dermalogical products and often replaces sodium azide as a biological buffer. It can also be found as a preservative in cosmetics, vaccines and various pharmaceuticals. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
IUPAC Name 2-phenoxyethan-1-ol
SMILES OCCOC1=CC=CC=C1

Specifications

CAS Max % 1
Boiling Point 244 to 250°C
Packaging Amber Glass Bottle
Quantity 120 mL
Percent Purity 99%
Grade Reagent
Physical Form Liquid