missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxyacetyl Chloride 98.0+%, TCI America™
$55.92 - $652.41
Chemical Identifiers
| CAS | 701-99-5 |
|---|---|
| Molecular Formula | C8H7ClO2 |
| Molecular Weight (g/mol) | 170.59 |
| MDL Number | MFCD00000726 |
| InChI Key | PKUPAJQAJXVUEK-UHFFFAOYSA-N |
| Synonym | phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride |
| PubChem CID | 69703 |
| IUPAC Name | 2-phenoxyacetyl chloride |
| SMILES | ClC(=O)COC1=CC=CC=C1 |
Chemical Identifiers
| 701-99-5 | |
| 170.59 | |
| PKUPAJQAJXVUEK-UHFFFAOYSA-N | |
| 69703 | |
| ClC(=O)COC1=CC=CC=C1 |
| C8H7ClO2 | |
| MFCD00000726 | |
| phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride | |
| 2-phenoxyacetyl chloride |
Specifications
| 701-99-5 | |
| 226°C | |
| MFCD00000726 | |
| 3265 | |
| PKUPAJQAJXVUEK-UHFFFAOYSA-N | |
| 2-phenoxyacetyl chloride | |
| 69703 | |
| ≥98.0% (GC,T) | |
| Phenoxyacetyl Chloride |
| Yellow | |
| C8H7ClO2 | |
| 25 g | |
| phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride | |
| ClC(=O)COC1=CC=CC=C1 | |
| 170.59 | |
| 170.59 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-1714&(9)-1262
TSCA : Yes