accessibility menu, dialog, popup

UserName

CAS RN 701-99-5

IUPAC Name: 2-phenoxyacetyl chloride
Synonyms: 2-phenoxyacetyl chloride
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: PKUPAJQAJXVUEK-UHFFFAOYSA-N
SMILES: ClC(=O)COC1=CC=CC=C1
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)