missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetyl Chloride 98.0+%, TCI America™

Catalog No. P011325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011325G 25 g
P0113500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P011325G Supplier TCI America Supplier No. P011325G

Chemical Identifiers

CAS 701-99-5
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000726
InChI Key PKUPAJQAJXVUEK-UHFFFAOYSA-N
Synonym phenoxyacetyl chloride, acetyl chloride, phenoxy, phenyloxyacetyl chloride, phenoxyacetylchloride, phenoxyacetic acid chloride, acetyl chloride, 2-phenoxy, pac-cl, phenoxylacetylchloride, 2-phenoxyacetylchloride, phenoxy acetyl chloride
PubChem CID 69703
IUPAC Name 2-phenoxyacetyl chloride
SMILES ClC(=O)COC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 226°C
Quantity 25 g
UN Number 3265
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Phenoxyacetyl Chloride
EINECSNumber (3)-1714&(9)-1262
TSCA Yes