missing translation for 'onlineSavingsMsg'
Learn More

Phenoxyacetic Acid 98.0+%, TCI America™

Catalog No. P0107500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0107500G 500 g
P010725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0107500G Supplier TCI America Supplier No. P0107500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122-59-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00004296
InChI Key LCPDWSOZIOUXRV-UHFFFAOYSA-N
Synonym phenoxyacetic acid, phenoxyethanoic acid, acetic acid, phenoxy, glycolic acid phenyl ether, phenoxy acetic acid, glycol acid phenyl ether, o-phenylglycolic acid, phenoxyacetate, glycollic acid phenyl ether, acide phenoxyacetique
PubChem CID 19188
ChEBI CHEBI:8075
IUPAC Name 2-phenoxyacetic acid
SMILES OC(=O)COC1=CC=CC=C1

Specifications

Melting Point 100°C
Boiling Point 285°C
Quantity 500 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Phenoxyacetic Acid
EINECSNumber (9)-1261
RTECSNumber AJ2230000
TSCA Yes