missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxyacetic acid, 98+%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 98.0°C to 101.0°C |
| Color | White |
| Boiling Point | 285.0°C |
| Flash Point | 165°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (Acid) |
| Packaging | Plastic bottle |
| Linear Formula | C6H5OCH2CO2H |
| Quantity | 500 g |
| Beilstein | 06,161 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Wear protective gloves/protective cloth |
| EINECSNumber | 204-556-7 |
RUO – Research Use Only