Learn More
Phenothiazine, 98+%
CAS: 92-84-2 | C12H9NS | 199.271 g/mol
Supplier: Thermo Scientific Chemicals A1251736
Description
It acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt acts on type I calcium/calmodulin dependent phosphodiesterase and also a useful electron donor. Phenothiazine finds uses in metal free organic dye sensitizers, dyes and antioxidants.
Solubility
Soluble in water (0.13 mg/ml at 23°C), ethanol, acetone, and ether. Insoluble in chloroform.
Notes
Store under dry inert gas. Protect from sunlight. Ensure good ventilation.
Specifications
| Phenothiazine | |
| 182°C to 186°C | |
| C12H9NS | |
| 500 g | |
| Light sensitive | |
| phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazine | |
| WJFKNYWRSNBZNX-UHFFFAOYSA-N | |
| 10H-phenothiazine | |
| 7108 | |
| 199.28 |
| 92-84-2 | |
| 235°C (21 mmHg) | |
| MFCD00005015 | |
| 143237 | |
| 14,7252 | |
| Soluble in water (0.13mg/ml at 23°C),ethanol,acetone,and ether. Insoluble in chloroform. | |
| C1=CC=C2C(=C1)NC3=CC=CC=C3S2 | |
| 199.271 | |
| CHEBI:37931 | |
| ≥98% |
Chemical Identifiers
| 92-84-2 | |
| 199.271 | |
| WJFKNYWRSNBZNX-UHFFFAOYSA-N | |
| 7108 | |
| 10H-phenothiazine |
| C12H9NS | |
| MFCD00005015 | |
| phenothiazine, thiodiphenylamine, feeno, dibenzothiazine, phenosan, phenthiazine, dibenzo-1,4-thiazine, penthazine, souframine, agrazine | |
| CHEBI:37931 | |
| C1=CC=C2C(=C1)NC3=CC=CC=C3S2 |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
H302-H317-H373
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 202-196-5
RTECSNumber : SN5075000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only