accessibility menu, dialog, popup

UserName

CAS RN 92-84-2

IUPAC Name: 10H-phenothiazine
Synonyms: phenothiazine phenoverm caswell no. 652 agrazine nexarbol phenthiazinum fentiazin antiverm thiodifenylamine wurm-thional
Molecular Weight (g/mol): 199.27
Molecular Formula: C12H9NS
InChi Key: WJFKNYWRSNBZNX-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2SC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)