missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Solution, Alcoholic, 1.0%, Fisher Chemical™

Catalog No. SP62500
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SP62500 500 mL Glass Bottle
SP621 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SP62500 Supplier Thermo Fisher Scientific Supplier No. FLSP62500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Volumetric indicator

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Chemical Identifiers

CAS 77-09-8 , 67-63-0
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point -89°C
Boiling Point 83°C
Concentration 0.7 to 1.3%
Packaging Glass Bottle
Vapor Pressure 40mmHg
Visual Transition Interval Pass Test
Density 0.7855g/cm³
Color Colorless
Quantity 500 mL
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 1% w/v
Chemical Name or Material Phenolphthalein Solution, Alcoholic 1.0%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid