missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
CAS: 62625-16-5 | C20H13K3O11S2 | 610.73 g/mol
$364.29 - $571.73
Chemical Identifiers
| CAS | 62625-16-5 |
|---|---|
| Molecular Formula | C20H13K3O11S2 |
| Molecular Weight (g/mol) | 610.73 |
| MDL Number | MFCD00150194 |
| InChI Key | PLHSVCWOEHYSHO-UHFFFAOYSA-K |
| PubChem CID | 129893851 |
| SMILES | [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC417220050
|
Thermo Scientific Chemicals
417220050 |
5 g | Glass bottle |
Each for $364.29
|
|
||||
|
AC417220100
|
Thermo Scientific Chemicals
417220100 |
10 g | Glass bottle |
Each for $571.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 62625-16-5 | |
| 610.73 | |
| PLHSVCWOEHYSHO-UHFFFAOYSA-K | |
| [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O |
| C20H13K3O11S2 | |
| MFCD00150194 | |
| 129893851 |
Specifications
| 230.0°C | |
| 98% | |
| MFCD00150194 | |
| PLHSVCWOEHYSHO-UHFFFAOYSA-K | |
| [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O | |
| 610.73 | |
| 664.78 | |
| Pure | |
| White | |
| Crystalline Powder |
| 62625-16-5 | |
| C20H13K3O11S2 | |
| Solubility in water: slightly soluble. | |
| Authentic | |
| tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate | |
| 129893851 | |
| 98% | |
| Glass bottle | |
| 5 g | |
| Phenolphthalein disulfate, tripotassium salt trihydrate |
Safety and Handling
EINECSNumber : 263-648-5
TSCA : TSCA
RUO – Research Use Only