missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure

Catalog No. AC417220050
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417220050 5 g Glass bottle
AC417220100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417220050 Supplier Thermo Scientific Chemicals Supplier No. 417220050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 62625-16-5 | C20H13K3O11S2 | 610.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 62625-16-5
Molecular Formula C20H13K3O11S2
Molecular Weight (g/mol) 610.73
MDL Number MFCD00150194
InChI Key PLHSVCWOEHYSHO-UHFFFAOYSA-K
PubChem CID 129893851
SMILES [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O

Specifications

Melting Point 230.0°C
Assay Percent Range 98%
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
IUPAC Name tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Formula Weight 664.78
Percent Purity 98%
Grade Pure
Packaging Glass bottle
Color White
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein disulfate, tripotassium salt trihydrate
Show More Show Less
EINECSNumber 263-648-5
TSCA TSCA

RUO – Research Use Only