missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Disulfate Potassium Salt Hydrate 97.0+%, TCI America™

Catalog No. P00971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00971G 1 g
P00975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P00971G Supplier TCI America Supplier No. P00971G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62625-16-5
Molecular Formula C20H13K3O11S2
Molecular Weight (g/mol) 610.73
MDL Number MFCD00150194
InChI Key PLHSVCWOEHYSHO-UHFFFAOYSA-K
Synonym phenolphthalein disulfate potassium salt, phenolphthalein disulfate tripotassium salt hydrate
PubChem CID 129893851
IUPAC Name tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
SMILES [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O

Specifications

Quantity 1 g
Formula Weight 610.73
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein Disulfate Potassium Salt Hydrate