missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 1% (w/v) in 60% (v/v) Isopropyl Alcohol, Neutralized, Ricca Chemical
Supplier: Ricca Chemical Company 5625616
Specifications
| Phenolphthalein | |
| 67-63-0,7732-18-5,77-09-8 | |
| 41.0,61.0,1.5 | |
| Indicator | |
| Natural Poly Bottle | |
| 8 | |
| Liquid |
| Neutralized | |
| 39.0,59.0,0.5 | |
| 1% W/V | |
| 1% (w/v) in 60% (v/v) Isopropyl Alcohol | |
| Colorless to Pink | |
| 500 mL |