Learn More
Thermo Scientific Chemicals Phenolphthalein, 0.5% w/v in alcohol
CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
$207.18 - $344.71
Chemical Identifiers
| CAS | 77-09-8 |
|---|---|
| Molecular Formula | C20H14O4 |
| Molecular Weight (g/mol) | 318.33 |
| MDL Number | MFCD00005913 |
| InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
| PubChem CID | 4764 |
| ChEBI | CHEBI:34914 |
| IUPAC Name | 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one |
| SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 77-09-8 | |
| 318.33 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| CHEBI:34914 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C20H14O4 | |
| MFCD00005913 | |
| 4764 | |
| 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one |
Specifications
| 77-09-8 | |
| MFCD00005913 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 318.33 | |
| CHEBI:34914 | |
| Alcohol-like | |
| Colorless | |
| 11°C (52°F) | |
| Phenolphthalein |
| C20H14O4 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one | |
| 4764 | |
| 318.33 | |
| 0.918 g/cm3 at 20°C | |
| 500 mL | |
| Liquid |
Safety and Handling
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H225-H302+H332-H319-H350-H371
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 201-004-7
TSCA : Yes
Recommended Storage : Ambient temperatures
For Manufacturing and Laboratory Use. Not for beverage use