missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein, 0.5% w/v in alcohol

CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

Supplier:  Thermo Scientific Chemicals 038703AP

 View more versions of this product

Catalog No. AA38703AP

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenolphthalein
C20H14O4
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.33
0.918 g/cm3 at 20°C
500 mL
Liquid
77-09-8
MFCD00005913
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
Alcohol-like
Colorless
11°C (52°F)

Chemical Identifiers

77-09-8
318.33
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302+H332-H319-H350-H371

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 201-004-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

For Manufacturing and Laboratory Use. Not for beverage use