Learn More
Fisherbrand™ Phenolic Standing Height Cabinet
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
 View more versions of this product
Catalog No. FBPH10636W
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Rust, rot, mold resistant construction.
  • Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
  • Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
  • All colors have a solid black core
Specifications
Specifications
2 years
22 in.
34.875 in.
55.9 cm
1
2
Phenolic
2
88.6 cm
36 in.
91.4 cm
Standing Height
White
Product Suggestions
Product Suggestions