Learn More
Fisherbrand™ Phenolic Standing Height Cabinet
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
Description
- Rust, rot, mold resistant construction.
- Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
- Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
- All colors have a solid black core
Specifications
| 36 in. | |
| 22 in. | |
| 55.9 cm | |
| 34.875 in. | |
| Standing Height | |
| Platinum |
| 2 years | |
| 88.6 cm | |
| 91.4 cm | |
| 1 | |
| 2 | |
| Phenolic |