missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, TCI America™

Supplier:  TCI America P010025G

 View more versions of this product

Catalog No. P010025G


Only null left

Description

Description

Specifications

Specifications

Phenol Red
C19H14O5S
Phenolsulfonphthalein
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
25 g
143-74-8
MFCD00003552
BELBBZDIHDAJOR-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
4766
354.38
Crystal/Powder

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
C19H14O5S
MFCD00003552
Phenolsulfonphthalein
CHEBI:31991
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0271

RTECSNumber : SJ7490000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions