missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, TCI America™

$33.10 - $99.27

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P01001G
View Documents
TCI America
P01001G
1 g N/A
Only null left
 
P010025G
View Documents
TCI America
P010025G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
C19H14O5S
MFCD00003552
Phenolsulfonphthalein
CHEBI:31991
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

143-74-8
MFCD00003552
BELBBZDIHDAJOR-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
4766
354.38
Crystal/Powder
C19H14O5S
Phenolsulfonphthalein
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
1 g
Phenol Red
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0271

RTECSNumber : SJ7490000

TSCA : Yes

SDS
Documents

Documents