missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, pure, indicator

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol

$53.88 - $325.00

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonephthalein
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151430050
SDS View Documents
Thermo Scientific Chemicals
151430050
5 g Glass bottle
Each for $53.88
Only null left
 
AC151430250
SDS View Documents
Thermo Scientific Chemicals
151430250
25 g Glass bottle
Each for $151.60
Only null left
 
AC151431000
SDS View Documents
Thermo Scientific Chemicals
151431000
100 g Glass bottle
Each for $325.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C19H14O5S
MFCD00003552
Phenolsulfonephthalein
CHEBI:31991

Specifications

>300.0°C
C19H14O5S
19, 91
Phenolsulfonephthalein
BELBBZDIHDAJOR-UHFFFAOYSA-N
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
Pure
1700 (558nm, in buffer) (E (1% min./1cm))
Beige/Brown to Gray
Crystalline Powder, Crystals, Flakes and/or Chunks
143-74-8
MFCD00003552
15, 7360
Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Authentic
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
4766
354.38
Glass bottle
from pH 6.8 (yellow) to pH 8.2 (red)
5 g
Phenol Red
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-609-7

RTECSNumber : SJ7490000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only