missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, pure, indicator

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol

Supplier:  Thermo Scientific Chemicals 151430250

 View more versions of this product

Catalog No. AC151430250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Phenol Red
143-74-8
MFCD00003552
15, 7360
Solubility in water: 0.77g/L. Other solubilities: soluble in ethanol
Authentic
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
4766
354.38
Glass bottle
from pH 6.8 (yellow) to pH 8.2 (red)
25 g
>300.0°C
C19H14O5S
19, 91
Phenolsulfonephthalein
BELBBZDIHDAJOR-UHFFFAOYSA-N
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
Pure
1700 (558nm, in buffer) (E (1% min./1cm))
Beige/Brown to Gray
Crystalline Powder, Crystals, Flakes and/or Chunks

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C19H14O5S
MFCD00003552
Phenolsulfonephthalein
CHEBI:31991
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-609-7

RTECSNumber : SJ7490000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only