missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 1% (w/v), Ricca Chemical

$89.16 - $234.94

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) Mixture
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5725-4
View Documents
Ricca Chemical Company
RI57254
120 mL Amber Glass Bottle
Each for $89.16
Only null left
 
R57250001A
View Documents
Ricca Chemical Company
R57250001A
1 L Natural Poly Bottle
Each for $234.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

34487-61-1
Mixture
23686673
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
C19H13NaO5S
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate

Specifications

34487-61-1,7732-18-5
100.0,1.01
C19H13NaO5S
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
Mixture
1% W/V
Amber Glass Bottle
Orange/Red
120 mL
Phenol Red Indicator
97.0,0.97
1% (w/v)
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
23686673
Indicator
Aqueous Solution
6 to 7.3
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'