missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 1% (w/v), Ricca Chemical

Catalog No. 57254
Change view
Click to view available options
Quantity:
120 mL
1 L
Packaging:
Amber Glass Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
57254 120 mL Amber Glass Bottle
R57250001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57254 Supplier Ricca Chemical Company Supplier No. RI57254

Chemical Identifiers

CAS 34487-61-1 , 7732-18-5
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) Mixture
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Indicator
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
Color Orange/Red
pH 6 to 7.3
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Phenol Red Indicator
Show More Show Less