missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 0.5% (w/v), Ricca Chemical

Catalog No. R5724500500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5724500500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5724500500 Supplier Ricca Chemical Company Supplier No. R5724500500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34487-61-1 , 7732-18-5
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

CAS Min % 98.0, 0.49
CAS Max % 100.0, 0.51
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Color Orange-Red
Quantity 500 mL
Chemical Name or Material Phenol Red Indicator
ShelfLife 24 months