missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 0.1% (w/v), Ricca Chemical

Catalog No. R5720100500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5720100500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R5720100500 Supplier Ricca Chemical Company Supplier No. R5720100500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34487-61-1 , 7732-18-5
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) Mixture
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Concentration 0.1% (w/v)
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Orange/Red
pH 6 to 7.3
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.1% W/V
Grade Indicator
Chemical Name or Material Phenol Red Indicator
Show More Show Less