missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red Indicator, 0.02% (w/v), Ricca Chemical
$47.63 - $136.38
Specifications
| Chemical Name or Material | Phenol Red Indicator |
|---|---|
| CAS | 7732-18-5,34487-61-1 |
| Percent Purity | 0.02% W/V |
| Grade | Indicator |
| Concentration or Composition (by Analyte or Components) | 0.02% (w/v) Aqueous Solution |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
5700-4
|
Ricca Chemical Company
57004 |
120 mL | Natural Poly Bottle |
Each for $47.63
|
|
||||
|
5700-16
|
Ricca Chemical Company
570016 |
500 mL | Natural Poly Bottle |
Each for $76.53
|
|
||||
|
R57000001A
|
Ricca Chemical Company
R57000001A |
1 L | Natural Poly Bottle |
Each for $136.38
|
|
||||
Specifications
| Phenol Red Indicator | |
| 0.02% W/V | |
| 0.02% (w/v) Aqueous Solution | |
| Orange/Red |
| 7732-18-5,34487-61-1 | |
| Indicator | |
| Natural Poly Bottle | |
| 7.15 to 7.25 |