missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red Indicator, 0.02% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 57004
Specifications
| Phenol Red Indicator | |
| 0.02% W/V | |
| 0.02% (w/v) Aqueous Solution | |
| Orange/Red | |
| 120 mL |
| 7732-18-5,34487-61-1 | |
| Indicator | |
| Natural Poly Bottle | |
| 7.15 to 7.25 |