missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$312.32 - $2942.88
Chemical Identifiers
| CAS | 143-74-8 |
|---|---|
| Molecular Formula | C19H14O5S |
| Molecular Weight (g/mol) | 354.38 |
| MDL Number | MFCD00003552 |
| InChI Key | BELBBZDIHDAJOR-UHFFFAOYSA-N |
| PubChem CID | 4766 |
| ChEBI | CHEBI:31991 |
| IUPAC Name | 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
| SMILES | OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Chemical Identifiers
| 143-74-8 | |
| 354.38 | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| CHEBI:31991 | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C19H14O5S | |
| MFCD00003552 | |
| 4766 | |
| 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol |
Specifications
| 143-74-8 | |
| MFCD00003552 | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 354.38 | |
| CHEBI:31991 | |
| BAKER ANALYZED™ ACS Reagent | |
| 5 g |
| C19H14O5S | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 4766 | |
| 354.39 | |
| Wide Mouth Amber Glass Bottle | |
| Phenol Red |