missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Specifications
Specifications
| Packaging | Wide Mouth Amber Glass Bottle |
| Quantity | 5 g |
| Formula Weight | 354.39 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Chemical Name or Material | Phenol Red |