missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, 0.02%, Aqueous, pH 6.8 to 8.4 Yellow to Red, Certified, LabChem™

Catalog No. LC182507
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC182507 125 mL
LC182501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC182507 Supplier LabChem Supplier No. LC182507

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34487-61-1 , 7732-18-5
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

CAS Max % 99.98, 0.02
Identification Passes Test
Packaging Poly Bottle
Linear Formula C19H13O5SNa
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Color Red
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 376.36
Grade Certified
Chemical Name or Material Phenol Red, 0.02%, Aqueous, pH 6.8 to 8.4 Yellow to Red
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature