missing translation for 'onlineSavingsMsg'
Learn More

Phenol, Loose Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7157972
Change view
Click to view available options
Quantity:
125 g
500 g
1 kg
6 x 125 g
6 x 500 g
50 lb.
4 x 1 kg
45 kg
Packaging:
Amber Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605077 125 g Amber Glass Bottle
18605079 500 g Amber Glass Bottle
18605078 6 x 125 g Amber Glass Bottle
18605081 1 kg Amber Glass Bottle
18605080 6 x 500 g Amber Glass Bottle
18605082 4 x 1 kg Amber Glass Bottle
18605085 50 lb. Amber Glass Bottle
18605086 45 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18605077 Supplier Spectrum Chemical Mfg Cor Supplier No. P1060125GM
Only null left

P1060, 108-95-2, C6H6O

Spectrum™ Chemical Phenol, Loose Crystal, Reagent, ACS is a white crystalline, aromatic organic compound that is volatile. It's extracted from petroleum and mainly used as a synthetic building block to produce a wide range of pharmaceutical products and drugs such as aspirin and oral analgesics. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

CAS Max % 1
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle
Color Colorless to White
Quantity 125 g
Physical Form Loose Crystals
Chemical Name or Material Phenol