Learn More
Phenol chloroform isoamyl alcohol mixture, BioUltra, For molecular biology, 125:24:1, MilliporeSigma™ Supelco™
$188.25 - $656.75
Chemical Identifiers
MDL Number MFCD00133763
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019328
View Documents
Merck Emd Millipore
77619100ML
100 mL
Each for $188.25
Only null left
111018263
View Documents
Merck Emd Millipore
77619500ML
500 mL
Each for $656.75
Only null left
Description
Description
Specifications
Chemical Identifiers
MFCD00133763
Specifications
136112-00-0
4 to 5(Water-phase, after extraction with Water, 1:1)
100 mL
1.12 g/mL (at 20°C)
MFCD00133763
For molecular biology
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'