missing translation for 'onlineSavingsMsg'
Learn More

Phenol chloroform isoamyl alcohol mixture, BioUltra, For molecular biology, 125:24:1, MilliporeSigma™ Supelco™

Catalog No. 111019328
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019328 100 mL
111018263 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019328 Supplier Merck Emd Millipore Supplier No. 77619100ML

Chemical Identifiers

MDL Number MFCD00133763

Specifications

CAS 136112-00-0
Density 1.12 g/mL (at 20°C)
pH 4 to 5(Water-phase, after extraction with Water, 1:1)
Quantity 100 mL
Grade For molecular biology