missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 5.2

Catalog No. AAJ62336AE
Change view
Click to view available options
Quantity:
100 mL
400 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62336AE 100 mL
AAJ62336AN 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62336AE Supplier Thermo Scientific Chemicals Supplier No. J62336AE
Only null left

Phenol-Chloroform-Isoamyl alcohol mixture in the ratio of 25:24:1 (v/v/v), CAS # 136112-00-0, 67-66-3, 123-51-3, is a solution used in the purification of nucleic acids.

Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • This solution contains a mixture of pure chloroform, isoamyl alcohol and phenol saturated with Tris-EDTA, with a stabilized pH of 8.0

Applications

  • Phenol-Chloroform-Isoamyl alcohol mixture solution (25:24:1) is used in the extraction of nucleic acids, DNA and RNA
  • When nucleic acids are extracted with the Phenol-Chloroform-Isoamyl alcohol mixture solution, a clear component separation is achieved. The proteins present in the sample are denatured and removed (collected in the organic phase or in the interphase), while nucleic acids remain in the aqueous phase. The chloroform denatures proteins and facilitates the separation of the aqueous and organic phases, and the isoamyl alcohol reduces foaming during extraction.
  • It is also used to generate DNA minisatellite fingerprints

Chemical Identifiers

CAS 136112-00-0
Molecular Formula C12H19Cl3O2
Molecular Weight (g/mol) 301.632
MDL Number MFCD00133763
InChI Key ZYWFEOZQIUMEGL-UHFFFAOYSA-N
Synonym phenol-chloroform-isoamyl alcohol mixture, chloroform; isoamyl alcohol; phenol, phenol chloroform isoamyl alcohol, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 125:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, vetec tm reagent grade, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 49.5:49.5:1, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 5.2, with alkaline buffer, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 6.7, with alkaline buffer
PubChem CID 66587205
IUPAC Name chloroform;3-methylbutan-1-ol;phenol
SMILES CC(C)CCO.C1=CC=C(C=C1)O.C(Cl)(Cl)Cl

Specifications

Color Colorless
UN Number UN2922
Solubility Information Soluble in water.
pH 5.2
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln.
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H314-H341-H351-H373-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only