missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Stock Standard, 5.00 ± 0.05% (w/v), Ricca Chemical
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Methods | AOAC/APHA for Selenium, AOAC for Disinfectant Testing |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 1 L |