missing translation for 'onlineSavingsMsg'
Learn More

Pheniramine Maleate 98.0+%, TCI America™

Catalog No. P227125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P227125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P227125G Supplier TCI America Supplier No. P227125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132-20-7
Molecular Formula C20H24N2O4
Molecular Weight (g/mol) 356.422
MDL Number MFCD00079250
InChI Key SSOXZAQUVINQSA-DASCVMRKSA-N
PubChem CID 73416365
IUPAC Name (Z)-but-2-enedioic acid;(3S)-N,N-dimethyl-3-phenyl-3-pyridin-2-ylpropan-1-amine
SMILES CN(C)CCC(C1=CC=CC=C1)C2=CC=CC=N2.C(=CC(=O)O)C(=O)O

Specifications

Melting Point 108°C
Color White
Quantity 25 g
Formula Weight 356.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pheniramine Maleate
RTECSNumber UT0175000
TSCA Yes