accessibility menu, dialog, popup

UserName

CAS RN 132-20-7

IUPAC Name: (2Z)-but-2-enedioic acid; dimethyl[3-phenyl-3-(pyridin-2-yl)propyl]amine
Synonyms: maleic acid; pheniramine
Molecular Weight (g/mol): 356.42
Molecular Formula: C20H24N2O4
InChi Key: SSOXZAQUVINQSA-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CN(C)CCC(C1=CC=CC=C1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results