missing translation for 'onlineSavingsMsg'
Learn More

Phenetole 98.0+%, TCI America™

Catalog No. E004325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004325ML 25 mL
E0043500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004325ML Supplier TCI America Supplier No. E004325ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00009090
InChI Key DLRJIFUOBPOJNS-UHFFFAOYSA-N
Synonym phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
PubChem CID 7674
ChEBI CHEBI:67129
IUPAC Name ethoxybenzene
SMILES CCOC1=CC=CC=C1

Spécifications

Melting Point -30°C
Color Colorless
Boiling Point 173°C
Quantity 25 mL
UN Number 3271
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Phenetole
EINECSNumber (3)-0557
RTECSNumber SI7700000
TSCA Yes