missing translation for 'onlineSavingsMsg'
Learn More

Phenetole, 98+%

Catalog No. AC221492500
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221492500 250 mL Glass bottle
AC221495000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221492500 Supplier Thermo Scientific Chemicals Supplier No. 221492500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 103-73-1 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-73-1
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00009090
InChI Key DLRJIFUOBPOJNS-UHFFFAOYSA-N
Synonym phenetole, benzene, ethoxy, ethyl phenyl ether, phenetol, phenyl ethyl ether, phenoxyethane, a phenoxyethane, ether, ethyl phenyl, unii-rb8lu2c57f
PubChem CID 7674
ChEBI CHEBI:67129
IUPAC Name ethoxybenzene
SMILES CCOC1=CC=CC=C1

Specifications

Melting Point -30.0°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 169.0°C to 170.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5055 to 1.5075
Linear Formula C6H5OC2H5
Quantity 250 mL
Beilstein 06, 140
Specific Gravity 0.96
Merck Index 15, 7343
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and carbon tetrachloride
Formula Weight 122.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Phenetole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-139-7
RTECSNumber SI7700000
TSCA TSCA

RUO – Research Use Only