missing translation for 'onlineSavingsMsg'
Learn More

Phenethyl Salicylate 98.0+%, TCI America™

Catalog No. P216525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P216525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P216525G Supplier TCI America Supplier No. P216525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-22-9
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD00020036
InChI Key YNMSDIQQNIRGDP-UHFFFAOYSA-N
Synonym Salicylic Acid Phenethyl Ester, 2-Phenylethyl Salicylate, Salicylic Acid 2-Phenylethyl Ester, Phenethyl 2-Hydroxybenzoate, 2-Hydroxybenzoic Acid Phenethyl Ester, 2-Phenylethyl 2-Hydroxybenzoate, 2-Hydroxybenzoic Acid 2-Phenylethyl Ester
PubChem CID 62332
IUPAC Name 2-phenylethyl 2-hydroxybenzoate
SMILES OC1=CC=CC=C1C(=O)OCCC1=CC=CC=C1

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 370°C
Quantity 25 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Phenethyl Salicylate
EINECSNumber (3)-1570
TSCA Yes