missing translation for 'onlineSavingsMsg'
Learn More

Phenazine 99.0+%, TCI America™

Supplier:  TCI America P008225G

 View more versions of this product

Catalog No. P008225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenazine
176°C
C12H8N2
25 g
PCNDJXKNXGMECE-UHFFFAOYSA-N
phenazine
4757
180.21
Crystalline Powder
92-82-0
Yellow
MFCD00005023
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2
180.21
CHEBI:36674
≥99.0% (HPLC)

Chemical Identifiers

92-82-0
180.21
PCNDJXKNXGMECE-UHFFFAOYSA-N
4757
phenazine
C12H8N2
MFCD00005023
dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs
CHEBI:36674
C1=CC=C2C(=C1)N=C3C=CC=CC3=N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SG1360000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions