missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenazine 99.0+%, TCI America™
$44.73 - $270.29
Chemical Identifiers
| CAS | 92-82-0 |
|---|---|
| Molecular Formula | C12H8N2 |
| Molecular Weight (g/mol) | 180.21 |
| MDL Number | MFCD00005023 |
| InChI Key | PCNDJXKNXGMECE-UHFFFAOYSA-N |
| Synonym | dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs |
| PubChem CID | 4757 |
| ChEBI | CHEBI:36674 |
| IUPAC Name | phenazine |
| SMILES | C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 |
Chemical Identifiers
| 92-82-0 | |
| 180.21 | |
| PCNDJXKNXGMECE-UHFFFAOYSA-N | |
| 4757 | |
| phenazine |
| C12H8N2 | |
| MFCD00005023 | |
| dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
| CHEBI:36674 | |
| C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 |
Specifications
| 92-82-0 | |
| Yellow | |
| MFCD00005023 | |
| dibenzopyrazine, azophenylene, dibenzoparadiazine, acridizine, 9,10-diazaanthracene, dibenzo-p-diazine, unii-2jhr6k463w, phenazine, dibenzo b,e pyrazine, acmc-209rhs | |
| C1=CC=C2C(=C1)N=C3C=CC=CC3=N2 | |
| 180.21 | |
| CHEBI:36674 | |
| ≥99.0% (HPLC) | |
| Phenazine |
| 176°C | |
| C12H8N2 | |
| 1 g | |
| PCNDJXKNXGMECE-UHFFFAOYSA-N | |
| phenazine | |
| 4757 | |
| 180.21 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : SG1360000
TSCA : Yes